Learn More
Fine Science Tools Stevens Tenotomy Scissors, Stainless Steel, Curved, 11 cm
SDP
Catalog No. NC0647230
Encompass
Tip Shape: Curved/ Tips: Blunt-Blunt/ Cutting Edge: 21mm/ Alloy / Material: Stainless Steel/ Length: 11cm/ Serrated: No/
Catalog No. NC0647230 Supplier Fine Science Tools Supplier No. 1406711
Tip Shape: Curved/ Tips: Blunt-Blunt/ Cutting Edge: 21mm/ Alloy / Material: Stainless Steel/ Length: 11cm/ Serrated: No/