Learn More
Medchemexpress LLC HY-13423 50mg Medchemexpress, Tenovin-1 CAS:380315-80-0 Purity:>98%
SDP
Catalog No. 502081362
Encompass_Preferred
Medchemexpress, HY-13423 50mg Tenovin-1 CAS:380315-80-0 Tenovin-1, a p53 activator, protects p53 from MDM2-mediated degradation. Tenovin-1 acts through inhibition of the protein-deacetylating activities of SirT1 and SirT2. Tenovin-1 is also a dihydroorotate dehydrogenase (DHODH) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1362 Supplier Medchemexpress LLC Supplier No. HY1342350MG
Medchemexpress, HY-13423 50mg Tenovin-1 CAS:380315-80-0 Tenovin-1, a p53 activator, protects p53 from MDM2-mediated degradation. Tenovin-1 acts through inhibition of the protein-deacetylating activities of SirT1 and SirT2. Tenovin-1 is also a dihydroorotate dehydrogenase (DHODH) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.