Learn More
Medchemexpress LLC HY-15510B 5mg Medchemexpress, Tenovin-6 (Hydrochloride) CAS:1011301-29-3 Purity:>98%
SDP
Catalog No. 502030640
Encompass_Preferred
Medchemexpress, HY-15510B 5mg Tenovin-6 (Hydrochloride) CAS:1011301-29-3 Tenovin-6 Hydrochloride, an analog of Tenovin-1 (HY-13423), is an activator of p53 transcriptional activity. Tenovin-6 Hydrochloride inhibits the protein deacetylase activities of purified human SIRT1, SIRT2, and SIRT3 with IC50s of 21 μM, 10 μM, and 67 μM, respectively. Tenovin-6 Hydrochloride also inhibits dihydroorotate dehydrogenase (DHODH)[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0640 Supplier Medchemexpress LLC Supplier No. HY15510B5MG
Medchemexpress, HY-15510B 5mg Tenovin-6 (Hydrochloride) CAS:1011301-29-3 Tenovin-6 Hydrochloride, an analog of Tenovin-1 (HY-13423), is an activator of p53 transcriptional activity. Tenovin-6 Hydrochloride inhibits the protein deacetylase activities of purified human SIRT1, SIRT2, and SIRT3 with IC50s of 21 μM, 10 μM, and 67 μM, respectively. Tenovin-6 Hydrochloride also inhibits dihydroorotate dehydrogenase (DHODH)[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.