Learn More
Cayman Chemical Tenovin-6 (hydrochloride), Purity: ≥95%, CAS: 1011301-29-3, 10 mg
SDP
Catalog No. 502953227
Encompass_Preferred
A analog of tenovin-1; elevates p53 activity in MCF-7 cells at 10 µM and reduces growth of ARN8 melanoma xenograft tumors in SCID mice at a dose of 50 mg/kg
Catalog No. 50-295-3227 Supplier Cayman Chemical Supplier No. 1308610 MG
A analog of tenovin-1; elevates p53 activity in MCF-7 cells at 10 µM and reduces growth of ARN8 melanoma xenograft tumors in SCID mice at a dose of 50 mg/kg