Learn More
Hach Company Tensette Pipet, 0.1-1.0 mL
Supplier:  Hach Company 1970001
Encompass
Perform routine laboratory pipetting quickly, economically, and accurately with Hach's automatic TenSette Pipet. Two models dispense from 0.1 to 1 ml or from 1 to 10 ml in 0.1 or 1.0 ml increments, respectively. The pipet saves time, reduces glassware cleanup and assures accuracy. The 0.1 to 1.0 mL model is specifically designed for use with Hach Voluette and PourRite standards in Standard Additions accuracy checks. Set includes pipet, manual and 100 disposable tips.
Catalog No. 50-199-5179