Learn More
Hach Company Tensette Pipet, 1.0-10 mL
Supplier:  Hach Company 1970010
Encompass
Perform routine laboratory pipetting quickly, economically, and accurately with Hach's automatic Tensette Pipet. Two models dispense from 0.1 to 1 mL or from 1 to 10 mL in 0.1 or 1.0 mL increments, respectively. The pipet saves time, reduces glassware cleanup and assures accuracy. Set includes one pipet, a manual and 50 disposable tips.
Catalog No. 50-199-5181