Learn More
Cole-Parmer Tensioning Tool for 18 to 175 Pound Cable/Zip Ties, Aluminum
Catalog No. NC9475209 Shop All Cole-Parmer Products
Encompass_Preferred
Adjust tension from light to very tight using a knob at the base of this tool. No twisting— tool cuts tie automatically when preset tension is reached. Tension knob features lock to ensure repeatable tension, tie after tie. Shock-resistant tool absorbs all impact.
Catalog No. NC9475209 Supplier Cole-Parmer Supplier No. 0683000
Adjust tension from light to very tight using a knob at the base of this tool. No twisting— tool cuts tie automatically when preset tension is reached. Tension knob features lock to ensure repeatable tension, tie after tie. Shock-resistant tool absorbs all impact.