Learn More
Cole-Parmer Tensioning Tool for 18 to 175 Pound Cable/Zip Ties, Polycarbonate
Catalog No. NC9493010 Shop All Cole-Parmer Products
Encompass_Preferred
Lightweight, polycarbonate tensioning tool features dial-adjustable tension setting. Simply squeeze the handle to pull tie to a preset tension and cut.
Catalog No. NC9493010 Supplier Cole-Parmer Supplier No. 0683007
Lightweight, polycarbonate tensioning tool features dial-adjustable tension setting. Simply squeeze the handle to pull tie to a preset tension and cut.