Learn More
Chem-Impex International, Inc. TentaGel S RAM (0.2 - 0.8 meq/g, 90 µm) | MFCD03792613 | 5G
Supplier:  Chem-Impex International, Inc. 047755G
Encompass_Preferred
TentaGel S RAM (0.2 - 0.8 meq/g, 90 µm), MFCD03792613, 5G
Catalog No. 50-500-001