Learn More
Enzo Life Sciences Tentoxin (0.5mg). CAS: 28540-82-1
SDP
Catalog No. 502008749
Encompass_Preferred
Cyclic tetrapeptide. Inhibitor of chloroplast F1-ATPase (CF1). Does not inhibit the homologous enzymes from chloroplasts of insusceptible plant species or from bacteria or mitochondria. Alternative name: Cyclo-N-Methyl-L-alanyl-L-leucyl-N-methyl-(Z)-dihydrophenylalanylglycyl. Purity: ≥98% (HPLC, TLC). Solubility: Soluble in DMSO (10mg/ml) or methanol (10mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-8749 Supplier Enzo Life Sciences Supplier No. ALX350132MC05
Cyclic tetrapeptide. Inhibitor of chloroplast F1-ATPase (CF1). Does not inhibit the homologous enzymes from chloroplasts of insusceptible plant species or from bacteria or mitochondria. Alternative name: Cyclo-N-Methyl-L-alanyl-L-leucyl-N-methyl-(Z)-dihydrophenylalanylglycyl. Purity: ≥98% (HPLC, TLC). Solubility: Soluble in DMSO (10mg/ml) or methanol (10mg/ml). Long Term Storage: +4°C.