Learn More
Medchemexpress LLC Tenuazonic acid-d13 | 2714417-30-6 | 99.2% | 210.31 | C10H2D13NO3 | 1 MG

Supplier: Medchemexpress LLC HYW6539211MG
Tenuazonic acid-d13 is a deuterium-labeled analytical standard of tenuazonic acid supplied as a white to off-white solid for research applications. It is used primarily as an internal standard or tracer in mass spectrometry and quantitative analysis, and is provided with manufacturer COA and SDS documentation for quality and safety.
- Deuterium-labeled internal standard for mass spectrometry quantification
- Molecular formula C10H2D13NO3 and molecular weight 210.31
- High purity: 99.2%
- Appearance: white to off-white solid
- Storage: powder -20°C (stable for 3 years); in solvent -80°C (6 months), -20°C (1 month)
- COA and SDS available for batch-specific quality and handling information
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.