Learn More
Medchemexpress LLC Tenuazonic acid-d13 | 2714417-30-6 | 99.2% | 210.31 | C10H2D13NO3 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYW653921500UG
Encompass_Preferred
Tenuazonic acid-d13 is a deuterium-labeled isotopologue of tenuazonic acid supplied as an analytical standard for research and quantitative mass spectrometry. The labeled compound is intended for use as an internal standard or tracer in LC-MS/MS and other analytical workflows to improve accuracy in quantification and method validation.
  • Deuterium-labeled (d13) isotopologue for mass spectrometry
  • High purity (99.2%) suitable for analytical use
  • Intended as an internal standard or tracer for quantitative assays
  • Available in small, pre-weighed quantities for method development and validation
  • Molecular formula C10H2D13NO3 and molecular weight 210.31 for analytical calculations
Catalog No. 50-002-83937