Learn More
Abcam Tenuazonic acid, mycoto x in with antibiotic, antiviral and antineoplastic activity, 1MG
Catalog No. 502706191
Encompass_Preferred
MW 197.23 Da, Purity >96%. Mycotoxin with antibiotic, antiviral and antineoplastic activity. Inhibits protein synthesis in vitro..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6191 Supplier Abcam Supplier No. AB1427641MG
MW 197.23 Da, Purity >96%. Mycotoxin with antibiotic, antiviral and antineoplastic activity. Inhibits protein synthesis in vitro..
The product is subject to the following: Abcam Restricted Use Statement