Learn More
Cayman Chemical TenuazonIc AcIdcoppR salt 10mg
SDP
Catalog No. 502951475
Encompass_Preferred
A major Alternaria mycotoxins commonly found as a food contaminant (LD50 = 548 μg/egg in the chicken embryo toxicity assay) and linked to esophageal cancer in human populations at risk of high exposure; Mice fed 100 mg/kg/day orally for 10 months developed precancerous lesions on the esophageal mucosa
Catalog No. 50-295-1475 Supplier Cayman Chemical Supplier No. 1144310 MG
A major Alternaria mycotoxins commonly found as a food contaminant (LD50 = 548 μg/egg in the chicken embryo toxicity assay) and linked to esophageal cancer in human populations at risk of high exposure; Mice fed 100 mg/kg/day orally for 10 months developed precancerous lesions on the esophageal mucosa