Learn More
Medchemexpress LLC HY-N0702 5mg Medchemexpress, Tenuifolin CAS:20183-47-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN07025MG
Encompass_Preferred
Medchemexpress, HY-N0702 5mg Tenuifolin CAS:20183-47-5 Tenuifolin is a triterpene isolated from Polygala tenuifolia Willd, has neuroprotective effects. Tenuifolin reduces Aβ secretion by inhibiting β-secretase. Tenuifolin improves learning and memory in aged mice by decreasing AChE activity and has the potential for Alzheimer’s disease (AD) treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2812