Learn More
Medchemexpress LLC Tenuifoliside B | 139726-36-6 | 5 MG
SDP
Catalog No. 5000418208
Encompass_Preferred
Tenuifoliside B is a component isolated from Polygalae Radix. It inhibits potassium cyanide (KCN)-induced hypoxia and scopolamine-induced memory impairment. This compound demonstrates potential cognitive improvement and cerebral protective effects, and it has the potential to be an anti-AD lead compound.
  • Demonstrates potential cognitive improvement
  • Demonstrates cerebral protective effects
  • Has the potential to be an anti-AD lead compound
Catalog No. 50-004-18208 Supplier Medchemexpress LLC Supplier No. HYN42685MG
Tenuifoliside B is a component isolated from Polygalae Radix. It inhibits potassium cyanide (KCN)-induced hypoxia and scopolamine-induced memory impairment. This compound demonstrates potential cognitive improvement and cerebral protective effects, and it has the potential to be an anti-AD lead compound.
  • Demonstrates potential cognitive improvement
  • Demonstrates cerebral protective effects
  • Has the potential to be an anti-AD lead compound