Learn More
Medchemexpress LLC Tenuifoliside B | 139726-36-6 | MFCD00915287 | 99.2% | 668.60 g/mol | C30H36O17 | 10 MG
SDP
Catalog No. 5000289851
Encompass_Preferred
Tenuifoliside B is a natural glycoside isolated from Polygala tenuifolia and supplied as a high-purity solid for laboratory research. It is reported to have neuroprotective and cognition-enhancing effects in preclinical studies and is intended for research use only.
  • Natural glycoside isolated from Polygala tenuifolia
  • White to yellow solid powder suitable for analytical and biological studies
  • Molecular formula C30H36O17 and molecular weight 668.60 g/mol
  • High purity (approx. 99.2%) for research applications
  • Reported biological activities include inhibition of KCN-induced hypoxia and protection against scopolamine-induced memory impairment
  • For research use only; not for human or clinical use
Catalog No. 50-002-89851 Supplier Medchemexpress LLC Supplier No. HYN426810MG
Tenuifoliside B is a natural glycoside isolated from Polygala tenuifolia and supplied as a high-purity solid for laboratory research. It is reported to have neuroprotective and cognition-enhancing effects in preclinical studies and is intended for research use only.
  • Natural glycoside isolated from Polygala tenuifolia
  • White to yellow solid powder suitable for analytical and biological studies
  • Molecular formula C30H36O17 and molecular weight 668.60 g/mol
  • High purity (approx. 99.2%) for research applications
  • Reported biological activities include inhibition of KCN-induced hypoxia and protection against scopolamine-induced memory impairment
  • For research use only; not for human or clinical use