Learn More
Medchemexpress LLC TEPC466 | 2621957-88-6 | 99.4% | 544.60 | 50 MG
SDP
Catalog No. 5000429707
Encompass_Preferred
TEPC466 is a novel TEPP-46-based aggregation-induced emission (AIE) probe that shows high selectivity and sensitivity for detecting the PKM2 protein via the AIE effect. It can be used for PKM2 detection and has been successfully applied in imaging the PKM2 protein in colorectal cancer cells with low toxicity. It is a useful tool for cancer diagnosis and research.
  • Novel TEPP-46-based aggregation-induced emission (AIE) probe
  • High degree of selectivity and sensitivity for the detection of PKM2 protein
  • Successfully applied in imaging PKM2 protein in colorectal cancer cells with low toxicity
  • Useful tool for cancer diagnosis and research
Catalog No. 50-004-29707 Supplier Medchemexpress LLC Supplier No. HYD145650MG
TEPC466 is a novel TEPP-46-based aggregation-induced emission (AIE) probe that shows high selectivity and sensitivity for detecting the PKM2 protein via the AIE effect. It can be used for PKM2 detection and has been successfully applied in imaging the PKM2 protein in colorectal cancer cells with low toxicity. It is a useful tool for cancer diagnosis and research.
  • Novel TEPP-46-based aggregation-induced emission (AIE) probe
  • High degree of selectivity and sensitivity for the detection of PKM2 protein
  • Successfully applied in imaging PKM2 protein in colorectal cancer cells with low toxicity
  • Useful tool for cancer diagnosis and research