Learn More
Medchemexpress LLC Tephrosin (Deguelinol I) | 76-80-2 | 410.42 | 1 MG
SDP
Catalog No. 5000415842
Encompass_Preferred
Tephrosin is a natural rotenoid exhibiting potent antitumor activities. It induces the degradation of EGFR and ErbB2 by promoting their internalization.
  • Potent antitumor activities
  • Induces degradation of EGFR
  • Induces degradation of ErbB2
  • Inhibits ligand-induced and constitutive phosphorylation of EGFR, ErbB2, and ErbB3
  • Suppresses activation of Akt and Erk1/2 mitogen-activated protein kinase (MAPK)
Catalog No. 50-004-15842 Supplier Medchemexpress LLC Supplier No. HYN11661MG
Tephrosin is a natural rotenoid exhibiting potent antitumor activities. It induces the degradation of EGFR and ErbB2 by promoting their internalization.
  • Potent antitumor activities
  • Induces degradation of EGFR
  • Induces degradation of ErbB2
  • Inhibits ligand-induced and constitutive phosphorylation of EGFR, ErbB2, and ErbB3
  • Suppresses activation of Akt and Erk1/2 mitogen-activated protein kinase (MAPK)