Learn More
Medchemexpress LLC Tepilamide fumarate | 1208229-58-6 | 99.8% | 243.26 | 100 MG
SDP
Catalog No. 5000346526
Encompass_Preferred
Tepilamide fumarate is an oral fumaric acid ester that acts as a proagent of Monomethyl fumarate. It is used in the research of moderate to severe chronic plaque psoriasis and can enhance the effectiveness of oncolytic viruses.
  • Oral fumaric acid ester
  • Proagent of Monomethyl fumarate
  • Enhances effectiveness of oncolytic viruses
  • Selectively promotes oncolytic virus infection in cancer cells without affecting healthy tissues
  • Used in research for moderate to severe chronic plaque psoriasis
  • Potential to enhance virus-based therapy
  • Solid, white to off-white appearance
Catalog No. 50-003-46526 Supplier Medchemexpress LLC Supplier No. HY10910550MG
Tepilamide fumarate is an oral fumaric acid ester that acts as a proagent of Monomethyl fumarate. It is used in the research of moderate to severe chronic plaque psoriasis and can enhance the effectiveness of oncolytic viruses.
  • Oral fumaric acid ester
  • Proagent of Monomethyl fumarate
  • Enhances effectiveness of oncolytic viruses
  • Selectively promotes oncolytic virus infection in cancer cells without affecting healthy tissues
  • Used in research for moderate to severe chronic plaque psoriasis
  • Potential to enhance virus-based therapy
  • Solid, white to off-white appearance