Learn More
Medchemexpress LLC Tepilamide fumarate | 1208229-58-6 | 99.8% | 243.26 | 50 MG
SDP
Catalog No. 5000344488
Encompass_Preferred
Tepilamide fumarate is an oral fumaric acid ester that acts as a proagent of Monomethyl fumarate. It is used in the research of moderate to severe chronic plaque psoriasis and can enhance the effectiveness of oncolytic viruses.
  • Acts as a proagent of Monomethyl fumarate
  • Used in research for moderate to severe chronic plaque psoriasis
  • Enhances the effectiveness of oncolytic viruses
  • Increases susceptibility of cancer cells to VSV∆51 infection
Catalog No. 50-003-44488 Supplier Medchemexpress LLC Supplier No. HY10910510MM/1ML
Tepilamide fumarate is an oral fumaric acid ester that acts as a proagent of Monomethyl fumarate. It is used in the research of moderate to severe chronic plaque psoriasis and can enhance the effectiveness of oncolytic viruses.
  • Acts as a proagent of Monomethyl fumarate
  • Used in research for moderate to severe chronic plaque psoriasis
  • Enhances the effectiveness of oncolytic viruses
  • Increases susceptibility of cancer cells to VSV∆51 infection