Learn More
Medchemexpress LLC HY-109105 5mg Medchemexpress, Tepilamide fumarate CAS:1208229-58-6 Purity:>98%
SDP
Catalog No. 501932341
Encompass_Preferred
Medchemexpress, HY-109105 5mg Tepilamide fumarate CAS:1208229-58-6 Tepilamide fumarate (XP-23829) is an oral fumaric acid ester, acts as a prodrug of monomethyl fumarate, and is used in the research of moderate to severe chronic plaque psoriasis [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2341 Supplier Medchemexpress LLC Supplier No. HY1091055MG
Medchemexpress, HY-109105 5mg Tepilamide fumarate CAS:1208229-58-6 Tepilamide fumarate (XP-23829) is an oral fumaric acid ester, acts as a prodrug of monomethyl fumarate, and is used in the research of moderate to severe chronic plaque psoriasis [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.