Learn More
Medchemexpress LLC Tepotinib | 1100598-32-0 | 99.9% | 492.57 | 5 MG
SDP
Catalog No. 501873072
Encompass_Preferred
Tepotinib (EMD-1214063) is an orally active, highly selective, reversible, ATP-competitive c-Met inhibitor. It is >200-fold selective for c-Met than IRAK4, TrkA, Axl, IRAK1, and Mer. Tepotinib inhibits c-Met phosphorylation, induces autophagy, and exhibits antitumor effects.
  • Highly selective c-Met inhibitor
  • Orally active
  • Reversible and ATP-competitive
  • Inhibits c-Met phosphorylation
  • Induces autophagy
  • Exhibits antitumor effects
Catalog No. 50-187-3072 Supplier Medchemexpress LLC Supplier No. HY147215MG
Tepotinib (EMD-1214063) is an orally active, highly selective, reversible, ATP-competitive c-Met inhibitor. It is >200-fold selective for c-Met than IRAK4, TrkA, Axl, IRAK1, and Mer. Tepotinib inhibits c-Met phosphorylation, induces autophagy, and exhibits antitumor effects.
  • Highly selective c-Met inhibitor
  • Orally active
  • Reversible and ATP-competitive
  • Inhibits c-Met phosphorylation
  • Induces autophagy
  • Exhibits antitumor effects