Learn More
Selleck Chemical LLC Tepotinib EMD 1214063-5mg
SDP
Supplier:  Selleck Chemical LLC S70675MG
Encompass_Preferred
Tepotinib is a potent and selective c-Met inhibitor with IC50 of 4 nM, >200-fold selective for c-Met than IRAK4, TrkA, Axl, IRAK1, and Mer. Tepotinib (EMD 1214063) induces autophagy. Phase 1.
Catalog No. 50-238-9882