Learn More
Medchemexpress LLC HY-18657 100mg Medchemexpress, TEPP-46 ML-265 CAS:1221186-53-3 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY18657100MG
Encompass_Preferred
Medchemexpress,HY-18657 100mg TEPP-46 ML-265 CAS:1221186-53-3 Formula:C17H16N4O2S2 TEPP-46 is a potent and selective pyruvate kinase M2 (PKM2) activator with an AC50 of 92 nM, showing little or no effect on PKM1, PKL and PKR. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8787