Learn More
TARGETMOL CHEMICALS INC TEPP-46 10MG
SDP
Catalog No. 502579238
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TEPP-46 (ML265) is a potent PKM2 activator (AC50 92 nM) showing little or no effect on PKM1 PKL and PKRinduces tetramerization and reduces tumor formation and size in a mouse xenograft model. . purity: 98%
Catalog No. 50-257-9238 Supplier TARGETMOL CHEMICALS INC Supplier No. T721410MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TEPP-46 (ML265) is a potent PKM2 activator (AC50 92 nM) showing little or no effect on PKM1 PKL and PKRinduces tetramerization and reduces tumor formation and size in a mouse xenograft model. . purity: 98%