Learn More
Medchemexpress LLC TEPP-46-based aggregation-induced emission (AIE) probe | 2621957-88-6 | 99.4% | 544.6 g/mol | C27H20N4O5S2 | 10 MG
SDP
Catalog No. 5000297799
Encompass_Preferred
TEPC466 is an aggregation-induced emission (AIE) fluorescent probe based on TEPP-46, designed for selective detection and imaging of pyruvate kinase M2 (PKM2) in biological samples. It is used for cellular imaging applications and is supplied as a high-purity research chemical.
  • Selective detection of PKM2 via the AIE effect.
  • Suitable for cellular imaging applications.
  • High purity (99.43%) for reliable experimental results.
  • Molecular weight 544.6 g/mol; formula C27H20N4O5S2.
  • Multiple pack sizes available for experimental flexibility.
  • Accompanied by analytical documents such as COA, HNMR, LCMS, and RP-HPLC.
Catalog No. 50-002-97799 Supplier Medchemexpress LLC Supplier No. HYD145610MG
TEPC466 is an aggregation-induced emission (AIE) fluorescent probe based on TEPP-46, designed for selective detection and imaging of pyruvate kinase M2 (PKM2) in biological samples. It is used for cellular imaging applications and is supplied as a high-purity research chemical.
  • Selective detection of PKM2 via the AIE effect.
  • Suitable for cellular imaging applications.
  • High purity (99.43%) for reliable experimental results.
  • Molecular weight 544.6 g/mol; formula C27H20N4O5S2.
  • Multiple pack sizes available for experimental flexibility.
  • Accompanied by analytical documents such as COA, HNMR, LCMS, and RP-HPLC.