Learn More
Medchemexpress LLC Teprenone | 3796-63-2 | >99.8% | 50 MG
SDP
Catalog No. 5000407856
Encompass_Preferred
Teprenone, also known as (5E,9E,13E)-Geranylgeranylacetone, is an isomer with antiulcer activity. It induces transcriptional activation of HSP genes, which may enhance gastric mucosal defense under stress conditions. This product is for research use only.
  • Isomer with antiulcer activity
  • Induces transcriptional activation of HSP genes
  • May increase gastric mucosal defense at conditions of stress
  • For research use only
Catalog No. 50-004-07856 Supplier Medchemexpress LLC Supplier No. HYB0779A50MG
Teprenone, also known as (5E,9E,13E)-Geranylgeranylacetone, is an isomer with antiulcer activity. It induces transcriptional activation of HSP genes, which may enhance gastric mucosal defense under stress conditions. This product is for research use only.
  • Isomer with antiulcer activity
  • Induces transcriptional activation of HSP genes
  • May increase gastric mucosal defense at conditions of stress
  • For research use only