Learn More
STEMCELL Technologies Anti-Mouse TER119 Antibody, Clone TER-119, Biotin, Size: 100 ug
SDP
Supplier:  STEMCELL Technologies 1000456
Encompass
Clone TER-119 is a rat (WI) monoclonal antibody that reacts with mouse TER119 (Ly-76) an ~52 kDa protein associated with glycophorin A on the surface of cells of the erythroid lineage in embryonic yolk sac fetal and newborn liver adult bone marrow peripheral blood and lymphoid organs. Verified for use with flow cytometry assays use this antibody in applications requiring the detection of TER119. The TER-119 antibody is a component of the "lineage cocktail" used to detect or deplete cells committed to hematopoietic lineages. In adult mice TER119 is found on ~20 - 25% of bone marrow cells and ~2 - 3% of splenocytes. This antibody clone has been verified for purity assessments of cells isolated with EasySep kits including EasySep Mouse CD4 T Cell Isolation Kit (Catalog 19852). Visit STEMCELL.com for more information and related products.
Catalog No. 50-239-0337