Learn More
Medchemexpress LLC Terazosin-d8 | 1006718-20-2 | 99.2% | 1 MG
SDP
Catalog No. 5000406630
Encompass_Preferred
Terazosin-d8 is a deuterium labeled version of Terazosin, a quinazoline derivative and a competitive, orally active α1-adrenoceptor antagonist. It functions by relaxing blood vessels and opening the bladder, showing potential for treating benign prostatic hyperplasia (BPH) and high blood pressure. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Competitive and orally active α1-adrenoceptor antagonist
  • Relaxing blood vessels and opening the bladder
  • Potential for benign prostatic hyperplasia (BPH) and high blood pressure treatment
Catalog No. 50-004-06630 Supplier Medchemexpress LLC Supplier No. HYB0371S1MG
Terazosin-d8 is a deuterium labeled version of Terazosin, a quinazoline derivative and a competitive, orally active α1-adrenoceptor antagonist. It functions by relaxing blood vessels and opening the bladder, showing potential for treating benign prostatic hyperplasia (BPH) and high blood pressure. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Competitive and orally active α1-adrenoceptor antagonist
  • Relaxing blood vessels and opening the bladder
  • Potential for benign prostatic hyperplasia (BPH) and high blood pressure treatment