Learn More
Medchemexpress LLC Terazosin-d8 | 1006718-20-2 | 99.2% | 2.5 MG
SDP
Catalog No. 5000406631
Encompass_Preferred
Terazosin-d8 is a deuterium-labeled quinazoline derivative and a competitive, orally active α1-adrenoceptor antagonist. It functions by relaxing blood vessels and opening the bladder, making it suitable for potential applications in treating benign prostatic hyperplasia (BPH) and high blood pressure. Deuteration influences the pharmacokinetic and metabolic profiles of drugs.
  • Competitive and orally active α1-adrenoceptor antagonist
  • Relaxes blood vessels and opens the bladder
  • Potential for benign prostatic hyperplasia (BPH) treatment
  • Potential for high blood pressure treatment
  • Can be used as a tracer or an internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06631 Supplier Medchemexpress LLC Supplier No. HYB0371S2.5MG
Add to Cart
Add to Cart
Terazosin-d8 is a deuterium-labeled quinazoline derivative and a competitive, orally active α1-adrenoceptor antagonist. It functions by relaxing blood vessels and opening the bladder, making it suitable for potential applications in treating benign prostatic hyperplasia (BPH) and high blood pressure. Deuteration influences the pharmacokinetic and metabolic profiles of drugs.
  • Competitive and orally active α1-adrenoceptor antagonist
  • Relaxes blood vessels and opens the bladder
  • Potential for benign prostatic hyperplasia (BPH) treatment
  • Potential for high blood pressure treatment
  • Can be used as a tracer or an internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS