Learn More
Apexbio Technology LLC Terazosin HCl 70024-40-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089346
Encompass_Preferred
Terazosin hydrochloride (CAS 70024-40-7) is a selective antagonist of alpha1-adrenergic receptors By inhibiting these receptors terazosin induces relaxation of smooth muscle in the prostate and bladder neck thereby reducing urinary outflow resistance It is widely utilized in biomedical research to study the modulation of alpha1-adrenergic signaling pathways and the pathophysiology of lower urinary tract symptoms particularly in the context of benign prostatic hyperplasia (BPH) Its mechanism supports investigations into autonomic regulation of smooth muscle function and the pharmacological management of urological disorders
Catalog No. 50-000-89346 Supplier Apexbio Technology LLC Supplier No. B13655.1
Terazosin hydrochloride (CAS 70024-40-7) is a selective antagonist of alpha1-adrenergic receptors By inhibiting these receptors terazosin induces relaxation of smooth muscle in the prostate and bladder neck thereby reducing urinary outflow resistance It is widely utilized in biomedical research to study the modulation of alpha1-adrenergic signaling pathways and the pathophysiology of lower urinary tract symptoms particularly in the context of benign prostatic hyperplasia (BPH) Its mechanism supports investigations into autonomic regulation of smooth muscle function and the pharmacological management of urological disorders