Learn More
Medchemexpress LLC HY-B0371A 100mg Medchemexpress, Terazosin (hydrochloride dihydrate) CAS:70024-40-7 Purity:>98%
SDP
Catalog No. 502082496
Encompass_Preferred
Medchemexpress, HY-B0371A 100mg Terazosin (hydrochloride dihydrate) CAS:70024-40-7 Terazosin hydrochloride dihydrate is a quinazoline derivative and a competitive and orally active α1-adrenoceptor antagonist. Terazosin hydrochloride dihydrate works by relaxing blood vessels and the opening of the bladder. Terazosin hydrochloride dihydrate has the potential for benign prostatic hyperplasia (BPH) and high blood pressure treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2496 Supplier Medchemexpress LLC Supplier No. HYB0371A100MG
Medchemexpress, HY-B0371A 100mg Terazosin (hydrochloride dihydrate) CAS:70024-40-7 Terazosin hydrochloride dihydrate is a quinazoline derivative and a competitive and orally active α1-adrenoceptor antagonist. Terazosin hydrochloride dihydrate works by relaxing blood vessels and the opening of the bladder. Terazosin hydrochloride dihydrate has the potential for benign prostatic hyperplasia (BPH) and high blood pressure treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.