Learn More
Medchemexpress LLC Terazosin hydrochloride | 63074-08-8 | 1 ML
SDP
Catalog No. 5000406629
Encompass_Preferred
Terazosin hydrochloride is a quinazoline derivative and a competitive, orally active α1-adrenoceptor antagonist. It functions by relaxing blood vessels and facilitating the opening of the bladder. This compound shows potential for the treatment of benign prostatic hyperplasia (BPH) and high blood pressure.
  • Functions by relaxing blood vessels
  • Facilitates the opening of the bladder
  • Potential for treating benign prostatic hyperplasia (BPH)
  • Potential for treating high blood pressure
Catalog No. 50-004-06629 Supplier Medchemexpress LLC Supplier No. HYB0371F10MM/1ML
Terazosin hydrochloride is a quinazoline derivative and a competitive, orally active α1-adrenoceptor antagonist. It functions by relaxing blood vessels and facilitating the opening of the bladder. This compound shows potential for the treatment of benign prostatic hyperplasia (BPH) and high blood pressure.
  • Functions by relaxing blood vessels
  • Facilitates the opening of the bladder
  • Potential for treating benign prostatic hyperplasia (BPH)
  • Potential for treating high blood pressure