Learn More
Medchemexpress LLC Terazosin hydrochloride | 63074-08-8 | 99.9% | 100 MG
SDP
Catalog No. 5000406632
Encompass_Preferred
Terazosin hydrochloride is a quinazoline derivative and a competitive and orally active α1-adrenoceptor antagonist. It works by relaxing blood vessels and opening the bladder. It has potential for benign prostatic hyperplasia (BPH) and high blood pressure treatment. This product is for research use only and not sold to patients.
  • Competitive and orally active α1-adrenoceptor antagonist
  • Relaxes blood vessels and opens the bladder
  • Potential for benign prostatic hyperplasia (BPH) treatment
  • Potential for high blood pressure treatment
  • For research use only
Catalog No. 50-004-06632 Supplier Medchemexpress LLC Supplier No. HYB0371F100MG
Terazosin hydrochloride is a quinazoline derivative and a competitive and orally active α1-adrenoceptor antagonist. It works by relaxing blood vessels and opening the bladder. It has potential for benign prostatic hyperplasia (BPH) and high blood pressure treatment. This product is for research use only and not sold to patients.
  • Competitive and orally active α1-adrenoceptor antagonist
  • Relaxes blood vessels and opens the bladder
  • Potential for benign prostatic hyperplasia (BPH) treatment
  • Potential for high blood pressure treatment
  • For research use only