Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Terazosin hydrochloride | 63074-08-8 | 99.9% | 50 MG
SDP

Catalog No. 5000407140
Encompass_Preferred

Terazosin hydrochloride is a quinazoline derivative and a competitive and orally active α1-adrenoceptor antagonist. It functions by relaxing blood vessels and opening the bladder. It has potential for the treatment of benign prostatic hyperplasia (BPH) and high blood pressure.

  • Relaxes blood vessels
  • Opens the bladder
  • Potential for treatment of benign prostatic hyperplasia (BPH)
  • Potential for treatment of high blood pressure
  • For research use only
  • Solid appearance
  • White to off-white color
  • Purity: 99.9%
  • Molecular weight: 423.89
  • Molecular formula: C19H26ClN5O4

Catalog No. 50-004-07140 Supplier Medchemexpress LLC Supplier No. HYB0371F50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Terazosin hydrochloride is a quinazoline derivative and a competitive and orally active α1-adrenoceptor antagonist. It functions by relaxing blood vessels and opening the bladder. It has potential for the treatment of benign prostatic hyperplasia (BPH) and high blood pressure.

  • Relaxes blood vessels
  • Opens the bladder
  • Potential for treatment of benign prostatic hyperplasia (BPH)
  • Potential for treatment of high blood pressure
  • For research use only
  • Solid appearance
  • White to off-white color
  • Purity: 99.9%
  • Molecular weight: 423.89
  • Molecular formula: C19H26ClN5O4

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.