Learn More
Cayman Chemical Terazosin (hydrochloride), Purity: ≥98%, CAS: 63074-08-8, 250 mg
SDP
Catalog No. 502983756
Encompass_Preferred
A potent and selective α1-AR antagonist; selective for α1-AR (Kis = 3.28, 0.68, and 1.09 for α1A-, α1B- and α1D-ARs, respectively) over α2-AR subtypes (Kis = 1,510, 7.71, and 78.2 nM for A2a-, A2b-, and A2c-ARs, respectively); decreases blood pressure in spontaneously hypertensive rats when administered at 0.1-30 mg/kg
Catalog No. 50-298-3756 Supplier Cayman Chemical Supplier No. 20216250 MG
A potent and selective α1-AR antagonist; selective for α1-AR (Kis = 3.28, 0.68, and 1.09 for α1A-, α1B- and α1D-ARs, respectively) over α2-AR subtypes (Kis = 1,510, 7.71, and 78.2 nM for A2a-, A2b-, and A2c-ARs, respectively); decreases blood pressure in spontaneously hypertensive rats when administered at 0.1-30 mg/kg