Learn More
Apexbio Technology LLC Terbinafine 91161-71-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062399
Encompass_Preferred
Terbinafine is an allylamine-class antifungal that exerts fungicidal effects by selectively inhibiting fungal squalene epoxidase thereby disrupting ergosterol biosynthesis and altering fungal cell membrane integrity Research demonstrates Terbinafine s broad-spectrum activity against diverse dermatophytes molds and select dimorphic fungal pathogens with in vitro IC50 values generally ranging from approximately 0 001 to 0 06 g/mL dependent on fungal strain and experimental condition In biological and biomedical studies Terbinafine is frequently employed as a pharmacological tool to probe antifungal mechanisms screen fungal susceptibility profiles and investigate therapeutic strategies against fungal infections
Catalog No. 50-000-62399 Supplier Apexbio Technology LLC Supplier No. A85335.1
Terbinafine is an allylamine-class antifungal that exerts fungicidal effects by selectively inhibiting fungal squalene epoxidase thereby disrupting ergosterol biosynthesis and altering fungal cell membrane integrity Research demonstrates Terbinafine s broad-spectrum activity against diverse dermatophytes molds and select dimorphic fungal pathogens with in vitro IC50 values generally ranging from approximately 0 001 to 0 06 g/mL dependent on fungal strain and experimental condition In biological and biomedical studies Terbinafine is frequently employed as a pharmacological tool to probe antifungal mechanisms screen fungal susceptibility profiles and investigate therapeutic strategies against fungal infections