Learn More
Medchemexpress LLC Terbinafine | 91161-71-6 | MFCD00242672 | 99.96% | 291.43 | 500 MG
SDP
Catalog No. 5000385753
Encompass_Preferred
Terbinafine (TDT 067) is an orally active and potent antifungal agent. It is a potent non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. Terbinafine also demonstrates antibacterial activity against certain Gram-positive and Gram-negative bacteria. It is a click chemistry reagent that contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Orally active.
  • Potent antifungal agent.
  • Potent non-competitive inhibitor of squalene epoxidase.
  • Shows antibacterial activity against certain gram-positive and gram-negative bacteria.
  • Click chemistry reagent.
  • Contains an alkyne group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition with molecules containing azide groups.
Catalog No. 50-003-85753 Supplier Medchemexpress LLC Supplier No. HY17395A500MG
Terbinafine (TDT 067) is an orally active and potent antifungal agent. It is a potent non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. Terbinafine also demonstrates antibacterial activity against certain Gram-positive and Gram-negative bacteria. It is a click chemistry reagent that contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Orally active.
  • Potent antifungal agent.
  • Potent non-competitive inhibitor of squalene epoxidase.
  • Shows antibacterial activity against certain gram-positive and gram-negative bacteria.
  • Click chemistry reagent.
  • Contains an alkyne group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition with molecules containing azide groups.