Learn More
Apexbio Technology LLC Terbinafine HCl 78628-80-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094633
Encompass_Preferred
Terbinafine hydrochloride (CAS 78628-80-5) is a small-molecule inhibitor targeting squalene epoxidase It is designed to selectively inhibit this key enzyme within the fungal sterol biosynthesis pathway thereby disrupting cell membrane integrity Terbinafine hydrochloride exerts its biological activity primarily through non-competitive inhibition of squalene epoxidase In experimental studies Terbinafine hydrochloride demonstrates potent antifungal activity with reported IC50 values against fungal squalene epoxidase in the nanomolar range Based on these pharmacological properties Terbinafine hydrochloride holds research potential in antifungal drug mechanism studies fungal sterol metabolism and therapeutic strategies against fungal pathogens in experimental models
Catalog No. 50-000-94633 Supplier Apexbio Technology LLC Supplier No. B20475.1
Terbinafine hydrochloride (CAS 78628-80-5) is a small-molecule inhibitor targeting squalene epoxidase It is designed to selectively inhibit this key enzyme within the fungal sterol biosynthesis pathway thereby disrupting cell membrane integrity Terbinafine hydrochloride exerts its biological activity primarily through non-competitive inhibition of squalene epoxidase In experimental studies Terbinafine hydrochloride demonstrates potent antifungal activity with reported IC50 values against fungal squalene epoxidase in the nanomolar range Based on these pharmacological properties Terbinafine hydrochloride holds research potential in antifungal drug mechanism studies fungal sterol metabolism and therapeutic strategies against fungal pathogens in experimental models