Learn More
Cayman Chemical Terbinafine hydrochloride
Supplier: Cayman Chemical 100116191 G
A broad-spectrum antifungal agent; has activity against T. rubrum, T. metagrophytes, T. verrucosum, E. floccosum, M. canis, A. fumigatus, and S. schenckii (MIC50s = 0.003-0.8 μg/ml); selectively inhibits C. albicans squalene epoxidase over rat liver epoxidase (IC50s = 0.03 and 77 μM, respectively); induces cell cycle arrest at the G0/G1 phase in COLO 205 tumor cells and HUVECs from 90-120 μM