Learn More
Cayman Chemical Terbinafine (hydrochloride), Purity: ≥98%, CAS: 78628-80-5, 500 mg
SDP
Catalog No. 502888116
Encompass_Preferred
A broad-spectrum antifungal agent; has activity against T. rubrum, T. metagrophytes, T. verrucosum, E. floccosum, M. canis, A. fumigatus, and S. schenckii (MIC50s = 0.003-0.8 μg/ml); selectively inhibits C. albicans squalene epoxidase over rat liver epoxidase (IC50s = 0.03 and 77 μM, respectively); induces cell cycle arrest at the G0/G1 phase in COLO 205 tumor cells and HUVECs from 90-120 μM
Catalog No. 50-288-8116 Supplier Cayman Chemical Supplier No. 10011619500 MG
A broad-spectrum antifungal agent; has activity against T. rubrum, T. metagrophytes, T. verrucosum, E. floccosum, M. canis, A. fumigatus, and S. schenckii (MIC50s = 0.003-0.8 μg/ml); selectively inhibits C. albicans squalene epoxidase over rat liver epoxidase (IC50s = 0.03 and 77 μM, respectively); induces cell cycle arrest at the G0/G1 phase in COLO 205 tumor cells and HUVECs from 90-120 μM