Learn More
TARGETMOL CHEMICALS INC Terbinafine hydrochloride 1G
SDP
Catalog No. 502875984
Encompass_Preferred
Also available in 1 mL, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Terbinafine hydrochloride (KWD 2019) is a synthetic allylamine derivative structurally related to naftifine. Terbinafine is active against dermatophytes. Purity 99.89%
Catalog No. 50-287-5984 Supplier TARGETMOL CHEMICALS INC Supplier No. T14111G
Also available in 1 mL, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Terbinafine hydrochloride (KWD 2019) is a synthetic allylamine derivative structurally related to naftifine. Terbinafine is active against dermatophytes. Purity 99.89%