Learn More
Medchemexpress LLC Terbinafine hydrochloride | 78628-80-5 | 99.86% | 327.89 | 200 MG
SDP
Catalog No. 5000384059
Encompass_Preferred
Terbinafine hydrochloride (TDT 067 hydrochloride) is an orally active and potent antifungal agent. It is a potent non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. Terbinafine hydrochloride also shows antibacterial activity against certain Gram-positive and Gram-negative bacteria. It is a click chemistry reagent, containing an Alkyne group, and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Orally active and potent antifungal agent.
  • Potent non-competitive inhibitor of squalene epoxidase from Candida (Ki of 30 nM).
  • Shows antibacterial activity against certain Gram-positive and Gram-negative bacteria.
  • Click chemistry reagent.
Catalog No. 50-003-84059 Supplier Medchemexpress LLC Supplier No. HY17395200MG
Terbinafine hydrochloride (TDT 067 hydrochloride) is an orally active and potent antifungal agent. It is a potent non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. Terbinafine hydrochloride also shows antibacterial activity against certain Gram-positive and Gram-negative bacteria. It is a click chemistry reagent, containing an Alkyne group, and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Orally active and potent antifungal agent.
  • Potent non-competitive inhibitor of squalene epoxidase from Candida (Ki of 30 nM).
  • Shows antibacterial activity against certain Gram-positive and Gram-negative bacteria.
  • Click chemistry reagent.