Learn More
Medchemexpress LLC Terbinafine hydrochloride | 78628-80-5 | 99.9% | 327.89 | 1 G
SDP
Catalog No. 5000386042
Encompass_Preferred
Terbinafine hydrochloride is an orally active and potent antifungal agent. It is a potent non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. It also shows antibacterial activity against certain Gram-positive and Gram-negative bacteria. This compound is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Orally active and potent antifungal agent
  • Potent non-competitive inhibitor of squalene epoxidase from Candida (Ki: 30 nM)
  • Shows antibacterial activity against certain Gram-positive and Gram-negative bacteria
  • Click chemistry reagent with an Alkyne group
Catalog No. 50-003-86042 Supplier Medchemexpress LLC Supplier No. HY173951G
Terbinafine hydrochloride is an orally active and potent antifungal agent. It is a potent non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. It also shows antibacterial activity against certain Gram-positive and Gram-negative bacteria. This compound is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Orally active and potent antifungal agent
  • Potent non-competitive inhibitor of squalene epoxidase from Candida (Ki: 30 nM)
  • Shows antibacterial activity against certain Gram-positive and Gram-negative bacteria
  • Click chemistry reagent with an Alkyne group