Learn More
Medchemexpress LLC Terbinafine hydrochloride | 78628-80-5 | 99.9% | 327.89 | 1 ML
SDP
Catalog No. 5000384582
Encompass_Preferred
Terbinafine hydrochloride (TDT 067 hydrochloride) is an orally active and potent antifungal agent. It acts as a non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. This compound also exhibits antibacterial activity against certain Gram-positive and Gram-negative bacteria. Furthermore, it is a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • Primarily shows fungicidal action against most fungal pathogens, including dermatophytes, dimorphic, and filamentous fungi.
  • Specifically inhibits fungal ergosterol biosynthesis by blocking squalene epoxidation.
  • Leads to rapid accumulation of squalene and a deficiency of ergosterol in treated fungal cells.
  • Effective in experimental dermatophytoses following oral administration and topical application.
Catalog No. 50-003-84582 Supplier Medchemexpress LLC Supplier No. HY1739510MM/1ML
Terbinafine hydrochloride (TDT 067 hydrochloride) is an orally active and potent antifungal agent. It acts as a non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. This compound also exhibits antibacterial activity against certain Gram-positive and Gram-negative bacteria. Furthermore, it is a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • Primarily shows fungicidal action against most fungal pathogens, including dermatophytes, dimorphic, and filamentous fungi.
  • Specifically inhibits fungal ergosterol biosynthesis by blocking squalene epoxidation.
  • Leads to rapid accumulation of squalene and a deficiency of ergosterol in treated fungal cells.
  • Effective in experimental dermatophytoses following oral administration and topical application.