Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbium(III) chloride, anhydrous, 99.9% (REO)

Catalog No. AA4167803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4167803 1 g
AA4167806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4167803 Supplier Thermo Scientific Chemicals Supplier No. 04167803
Add to Cart

CAS: 10042-88-3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10042-88-3
Molecular Formula Cl3Tb
Molecular Weight (g/mol) 265.28
MDL Number MFCD00011258
InChI Key GFISHBQNVWAVFU-UHFFFAOYSA-K
IUPAC Name trichloroterbium
SMILES Cl[Tb](Cl)Cl

Specifications

Melting Point 588°C
Physical Form Powder
Quantity 1 g
Merck Index 14,9157
Solubility Information Soluble in water
Formula Weight 265.29
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Terbium(III) chloride, Anhydrous
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 233-132-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.