Learn More
Strem, An Ascensus Company CAS# 13798-24-8. 25g. Terbium(III) chloride hexahydrate (99.9%-Tb) (REO). MFCD00149870
SDP
Catalog No. 5090116083
Encompass
CAS# 13798-24-8. 25g. Terbium(III) chloride hexahydrate (99.9%-Tb) (REO). MFCD00149870. Molecular Weight: 265.29 (373.38). Molecular Formula: TbCl3∙6H2O. Color/form: white xtl. Strem# 93-6502. http://www.strem.com/catalog/v/93-6502/
Catalog No. 50-901-16083 Supplier Strem, An Ascensus Company Supplier No. 93650225G
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
CAS# 13798-24-8. 25g. Terbium(III) chloride hexahydrate (99.9%-Tb) (REO). MFCD00149870. Molecular Weight: 265.29 (373.38). Molecular Formula: TbCl3∙6H2O. Color/form: white xtl. Strem# 93-6502. http://www.strem.com/catalog/v/93-6502/