Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbium(III) chloride hexahydrate, REacton™, 99.99% (REO)

Catalog No. p-7048675
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1121009 10 g
AA1121018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1121009 Supplier Thermo Scientific Chemicals Supplier No. 01121009
Only null left
Add to Cart

CAS: 13798-24-8 | Cl3H12O6Tb | 373.37 g/mol

Terbium(III) chloride hexahydrate is used in the primary chemical intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13798-24-8
Molecular Formula Cl3H12O6Tb
Molecular Weight (g/mol) 373.37
MDL Number MFCD00149870
InChI Key ULJUVCOAZNLCJZ-UHFFFAOYSA-K
IUPAC Name terbium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 10 g
Merck Index 14,9157
Solubility Information soluble in water
Formula Weight 373.38 (265.29 Anhydrous)
Percent Purity 99.99%
Grade Reacton
Odor Odorless
Assay (REO)
Chemical Name or Material Terbium(III) chloride hexahydrate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 233-132-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.