Learn More
Terbium(III) chloride hexahydrate, REacton™, 99.99% (REO)
Description
Terbium(III) chloride hexahydrate is used in the primary chemical intermediates and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Crystalline Aggregates |
| Quantity | 50 g |
| Odor | Odorless |
| Merck Index | 14,9157 |
| Solubility Information | soluble in water |
| Formula Weight | 373.38 (265.29 Anhydrous) |
| Percent Purity | 99.99% |
| Grade | Reacton |
| Assay | (REO) |
| Chemical Name or Material | Terbium(III) chloride hexahydrate |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement P280-P305+P351+P338-P362-P321-P332+P313-P337+P313 Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse. Specific treatment (see ... on this label). If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. |
| EINECSNumber | 233-132-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.